![Pyrantel embonate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/128/076/53ad5477-a27b-4c74-a15b-45ab94d248b6/640/53ad5477-a27b-4c74-a15b-45ab94d248b6.png)
Pyrantel embonate
![Pyrantel embonate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/128/076/53ad5477-a27b-4c74-a15b-45ab94d248b6/640/53ad5477-a27b-4c74-a15b-45ab94d248b6.png)
Synonym(s):
Pyrantel pamoate, 1-Methyl-2-(2-[2-thienyl]ethenyl)-1,4,5,6-tetrahydropyrimidine compound with 4,4′-methylenebis(3-hydroxy-2-naphthoic acid), Pyrantel embonate
Linear Formula:
C11H14N2S · C23H16O6
CAS Number:
Molecular Weight:
594.68
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing