![Prosulfocarb certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/270/427/1e4d489f-34f6-4f5f-86a5-b823d895689c/640/1e4d489f-34f6-4f5f-86a5-b823d895689c.png)
Prosulfocarb
![Prosulfocarb certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/270/427/1e4d489f-34f6-4f5f-86a5-b823d895689c/640/1e4d489f-34f6-4f5f-86a5-b823d895689c.png)
Synonym(s):
S-Benzyl dipropylthiocarbamate
Empirical Formula (Hill Notation):
C14H21NOS
CAS Number:
Molecular Weight:
251.39
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2