![Propaquizafop PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/168/254/46afe5e0-beed-4eb9-b417-2979abee87cc/640/46afe5e0-beed-4eb9-b417-2979abee87cc.png)
Propaquizafop
![Propaquizafop PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/168/254/46afe5e0-beed-4eb9-b417-2979abee87cc/640/46afe5e0-beed-4eb9-b417-2979abee87cc.png)
Empirical Formula (Hill Notation):
C22H22ClN3O5
CAS Number:
Molecular Weight:
443.88
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2