Primeverin
Synonym(s):
4-Methoxy-2-[(6-O-β-D-xylopyranosyl-β-D-glucopyranosyl)oxy]benzoic acid methyl ester
Empirical Formula (Hill Notation):
C20H28O13
CAS Number:
Molecular Weight:
476.43
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing