![Pramipexole Dihydrochloride Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/219/316/86647dfb-2139-4124-acc3-5355b0f3644f/640/86647dfb-2139-4124-acc3-5355b0f3644f.jpg)
Pramipexole Dihydrochloride
![Pramipexole Dihydrochloride Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/219/316/86647dfb-2139-4124-acc3-5355b0f3644f/640/86647dfb-2139-4124-acc3-5355b0f3644f.jpg)
Synonym(s):
Pramipexole dihydrochloride monohydrate
Empirical Formula (Hill Notation):
C10H17N3S · 2HCl · H2O
CAS Number:
Molecular Weight:
302.26
Products
Brand
Formula Weight
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing