![Poly(ethylene glycol) (N-hydroxysuccinimide 5-pentanoate) ether 2-(biotinylamino)ethane average Mn 3,800](/deepweb/assets/sigmaaldrich/product/structures/513/992/94858464-6b5c-4514-94ed-52b67916c391/640/94858464-6b5c-4514-94ed-52b67916c391.png)
Poly(ethylene glycol) (N-hydroxysuccinimide 5-pentanoate) ether 2-(biotinylamino)ethane
![Poly(ethylene glycol) (N-hydroxysuccinimide 5-pentanoate) ether 2-(biotinylamino)ethane average Mn 3,800](/deepweb/assets/sigmaaldrich/product/structures/513/992/94858464-6b5c-4514-94ed-52b67916c391/640/94858464-6b5c-4514-94ed-52b67916c391.png)
Synonym(s):
Biotin polyoxyethylene maleimide, Biotin-PEG, Biotin-PEG maleimide, Biotinylated PEG, NHS - protected acid PEG, Poly(ethylene glycol) (N-hydroxysuccinimide acetic acid) ether biotin, biotin carboxylic acid PEG, protected valeric acid
Linear Formula:
C10H16N3O2S(C2H4O)nC8H8NO5
Products
Brand
Melting Point (°C)
Physical Form
Quality Segment
Functional Group
Functionality
Shape
Polymer Composition
Polymer Type
Reactivity
Reagent Type
Showing 1-1 of 1