Piperlongumine
Synonym(s):
5,6-Dihydro-1-(1-oxo-3-[3,4,5-trimethoxyphenyl]-trans-2-propenyl)-2[1H]-pyridinone, 5,6-Dihydro-1-[(2E)-1-oxo-3-(3,4,5-trimethoxyphenyl)-2-propen-1-yl]-2(1H)-Pyridinone, Piplartin, Piplartine, Piperlongumine
Empirical Formula (Hill Notation):
C17H19NO5
CAS Number:
Molecular Weight:
317.34
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Research Area
Quality Segment
Application
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing