![Photo-reactive Clickable trans-Sterol Probe 95%](/deepweb/assets/sigmaaldrich/product/structures/424/046/db656602-5a62-49c5-9566-08126a557578/640/db656602-5a62-49c5-9566-08126a557578.png)
Photo-reactive Clickable trans-Sterol Probe
![Photo-reactive Clickable trans-Sterol Probe 95%](/deepweb/assets/sigmaaldrich/product/structures/424/046/db656602-5a62-49c5-9566-08126a557578/640/db656602-5a62-49c5-9566-08126a557578.png)
Synonym(s):
(4R)-Hex-5-yn-1-yl 4-((3S,5S,10R,13R,17R)-3-hydroxy-10,13-dimethyl-1,2,3,4,5,7,8,9,10,11,12,13,14,15,16,17-hexadecahydrospiro[cyclopenta[α]phenanthrene-6,3′-diazirin]-17-yl)pentanoate
Empirical Formula (Hill Notation):
C30H46N2O3
Molecular Weight:
482.70
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing