![Phloroglucinol dihydrate analytical standard](/deepweb/assets/sigmaaldrich/product/structures/299/311/1e4f75f7-3f03-40fd-9ef0-6c08946fcc2e/640/1e4f75f7-3f03-40fd-9ef0-6c08946fcc2e.png)
Phloroglucinol dihydrate
![Phloroglucinol dihydrate analytical standard](/deepweb/assets/sigmaaldrich/product/structures/299/311/1e4f75f7-3f03-40fd-9ef0-6c08946fcc2e/640/1e4f75f7-3f03-40fd-9ef0-6c08946fcc2e.png)
Linear Formula:
C6H3(OH)3 · 2H2O
CAS Number:
Molecular Weight:
162.14
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing