Phenoxymethylpenicillin
Synonym(s):
Phenoxymethylpenicillinic acid, Penicillin V
Empirical Formula (Hill Notation):
C16H18N2O5S
CAS Number:
Molecular Weight:
350.39
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing