![Permethrin for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/195/465/1a92e588-c3af-43f0-a5b2-a2fa162bd422/640/1a92e588-c3af-43f0-a5b2-a2fa162bd422.png)
Permethrin for system suitability
![Permethrin for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/195/465/1a92e588-c3af-43f0-a5b2-a2fa162bd422/640/1a92e588-c3af-43f0-a5b2-a2fa162bd422.png)
Synonym(s):
Permethrin
Empirical Formula (Hill Notation):
C21H20Cl2O3
CAS Number:
Molecular Weight:
391.29
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing