Skip to Content
Merck
Parathyroid Hormone Fragment 1-34 bovine ≥97% (HPLC), powder

Parathyroid Hormone Fragment 1-34 bovine

Parathyroid Hormone Fragment 1-34 bovine ≥97% (HPLC), powder
Empirical Formula (Hill Notation):
C183H288N54O50S2
CAS Number:
Molecular Weight:
4108.71

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing