Skip to Content
Merck
Oxcarbazepine Related Compound C pharmaceutical secondary standard, certified reference material

Oxcarbazepine Related Compound C

Oxcarbazepine Related Compound C pharmaceutical secondary standard, certified reference material
Synonym(s):
Acridone, 9,10-Dihydro-9-oxoacridine
Empirical Formula (Hill Notation):
C13H9NO
CAS Number:
Molecular Weight:
195.22

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade