Skip to Content
Merck
Oxcarbazepine European Pharmacopoeia (EP) Reference Standard

Oxcarbazepine

Oxcarbazepine European Pharmacopoeia (EP) Reference Standard
Synonym(s):
10,11-Dihydro-10-oxo-5h-dibenz[b,f]azepine-5-carboxamide, Oxacarbazepine, Oxcarbazepine
Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique