Oseltamivir phosphate
Synonym(s):
(3R,4R,5S)-4-Acetamido-5-amino-3-(1-ethylpropoxy)-1-cyclohexene-1-carboxylic acid ethyl ester phosphate salt, Oseltamivir phosphate
Empirical Formula (Hill Notation):
C16H28N2O4 ·H3PO4
CAS Number:
Molecular Weight:
410.40
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing