Ononin
Synonym(s):
Formononetin 7-O-β-D-glucopyranoside
Empirical Formula (Hill Notation):
C22H22O9
CAS Number:
Molecular Weight:
430.40
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Format
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing