![Olanzapine Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/361/444/62b6cab3-bdb3-4e2d-b194-8430b8dcea94/640/62b6cab3-bdb3-4e2d-b194-8430b8dcea94.jpg)
Olanzapine Related Compound C
![Olanzapine Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/361/444/62b6cab3-bdb3-4e2d-b194-8430b8dcea94/640/62b6cab3-bdb3-4e2d-b194-8430b8dcea94.jpg)
Synonym(s):
Olanzapine N-oxide, 2-Methyl-4-(4-methylpiperazin-1-yl)-10H-benzo[b]thieno[2,3-e][1,4]diazepine 4′-N-oxide
Empirical Formula (Hill Notation):
C17H20N4OS
CAS Number:
Molecular Weight:
328.43
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2