Skip to Content
Merck
o-Cresol analytical standard

o-Cresol

o-Cresol analytical standard
Synonym(s):
2-Methylphenol, o-Cresol
Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing