Skip to Content
Merck
Ocimene mixture of isomers, stabilized, ≥90%

Ocimene

Ocimene mixture of isomers, stabilized, ≥90%
Synonym(s):
3,7-Dimethyl-1,3,6-octatrien, 3,7-Dimethyl-1,3,6-octatriene
Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Application
Organoleptic Class
Showing 1-1 of 1
Compare
Product Number
Description
Pricing