![N,N′-Diphenylthiourea analytical standard](/deepweb/assets/sigmaaldrich/product/structures/372/979/1c9c31a0-cd06-4982-9c3b-f5c26aa1d661/640/1c9c31a0-cd06-4982-9c3b-f5c26aa1d661.png)
N,N′-Diphenylthiourea
![N,N′-Diphenylthiourea analytical standard](/deepweb/assets/sigmaaldrich/product/structures/372/979/1c9c31a0-cd06-4982-9c3b-f5c26aa1d661/640/1c9c31a0-cd06-4982-9c3b-f5c26aa1d661.png)
Synonym(s):
1,3-Diphenyl-2-thiourea, Diphenylsulfourea, Thiocarbanilide, N,N′-Diphenylthiourea
Linear Formula:
(C6H5NH)2CS
CAS Number:
Molecular Weight:
228.31
Products
Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Purity
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing