Skip to Content
Merck
N-Methylcytisine phyproof® Reference Substance

N-Methylcytisine

N-Methylcytisine phyproof® Reference Substance
Synonym(s):
Caulophylline
Empirical Formula (Hill Notation):
C12H16N2O
CAS Number:
Molecular Weight:
204.27
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing