Skip to Content
Merck
vibrant-m

Nitroglycerin in propylene glycol

vibrant-m
Synonym(s):
Glycerol trinitrate solution, 1,2,3-Propanetriol 1,2,3-trinitrate solution
Empirical Formula (Hill Notation):
C3H5N3O9
Molecular Weight:
227.09

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1