![Nisin from Lactococcus lactis potency: ≥900 IU/mg](/deepweb/assets/sigmaaldrich/product/structures/200/611/4c3321f7-4ed0-4f16-a897-dabbc2bf4757/640/4c3321f7-4ed0-4f16-a897-dabbc2bf4757.png)
Nisin from Lactococcus lactis
![Nisin from Lactococcus lactis potency: ≥900 IU/mg](/deepweb/assets/sigmaaldrich/product/structures/200/611/4c3321f7-4ed0-4f16-a897-dabbc2bf4757/640/4c3321f7-4ed0-4f16-a897-dabbc2bf4757.png)
Empirical Formula (Hill Notation):
C143H230N42O37S7
CAS Number:
Molecular Weight:
3354.07
Products
Product Category
Brand
Formula Weight
Physical Form
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing