![Nickel(II) sulfate hexahydrate EMPLURA®](/deepweb/assets/sigmaaldrich/product/images/416/076/53566e58-8fe6-49fb-904e-a2cea87aa289/640/53566e58-8fe6-49fb-904e-a2cea87aa289.jpg)
Nickel(II) sulfate hexahydrate
![Nickel(II) sulfate hexahydrate EMPLURA®](/deepweb/assets/sigmaaldrich/product/images/416/076/53566e58-8fe6-49fb-904e-a2cea87aa289/640/53566e58-8fe6-49fb-904e-a2cea87aa289.jpg)
Synonym(s):
Nickel(II) sulfate hexahydrate
CAS Number:
Molecular Weight:
262.86
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing