Skip to Content
Merck
Nickel(II) 5,9,14,18,23,27,32,36-octabutoxy-2,3-naphthalocyanine Dye content 98 %

Nickel(II) 5,9,14,18,23,27,32,36-octabutoxy-2,3-naphthalocyanine

Nickel(II) 5,9,14,18,23,27,32,36-octabutoxy-2,3-naphthalocyanine Dye content 98 %
Synonym(s):
5,9,14,18,23,27,32,36-Octabutoxy-2,3-naphthalocyanine nickel(II) salt
Empirical Formula (Hill Notation):
C80H88N8NiO8
CAS Number:
Molecular Weight:
1348.30

Products

Brand
Formula Weight
Melting Point (°C)
Chemical Composition
Markush Class
Markush Group
Showing 1-1 of 1