Skip to Content
Merck
N-Bromosuccinimide ReagentPlus®, 99%

N-Bromosuccinimide

N-Bromosuccinimide ReagentPlus®, 99%
Synonym(s):
NBS, N-Bromosuccinimide
Empirical Formula (Hill Notation):
C4H4BrNO2
CAS Number:
Molecular Weight:
177.98

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing