![N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride](/deepweb/assets/sigmaaldrich/product/structures/754/667/4d896e65-b7f7-4967-8bf8-6ba6a93f4b0e/640/4d896e65-b7f7-4967-8bf8-6ba6a93f4b0e.png)
N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride
![N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide hydrochloride](/deepweb/assets/sigmaaldrich/product/structures/754/667/4d896e65-b7f7-4967-8bf8-6ba6a93f4b0e/640/4d896e65-b7f7-4967-8bf8-6ba6a93f4b0e.png)
Synonym(s):
W-7
Empirical Formula (Hill Notation):
C16H21ClN2O2S · HCl
CAS Number:
Molecular Weight:
377.33
Products
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing