Skip to Content
Merck
Myristoyl coenzyme A lithium salt ≥80.0%

Myristoyl coenzyme A lithium salt

Myristoyl coenzyme A lithium salt ≥80.0%
Synonym(s):
n-Tetradecanoyl Coenzyme A lithium salt
Empirical Formula (Hill Notation):
C35H62N7O17P3S · xLi+
CAS Number:
Molecular Weight:
977.89 (free acid basis)

Products

Product Category
Brand
Formula Weight
Purity
Showing 1-1 of 1