![Monotropein phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/253/272/54b1aaac-1b78-43f4-9dd7-6a434ae55922/640/54b1aaac-1b78-43f4-9dd7-6a434ae55922.png)
Monotropein
![Monotropein phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/253/272/54b1aaac-1b78-43f4-9dd7-6a434ae55922/640/54b1aaac-1b78-43f4-9dd7-6a434ae55922.png)
Synonym(s):
[1S-(1α, 4aα,7β,7aα)]-1-(β-D-Glucopyranosyloxy)-1,4a,7,7a-tetrahydro-7-hydroxy-7-(hydroxymethyl)cyclopenta[c]pyran-4-carboxylic acid
Empirical Formula (Hill Notation):
C16H22O11
CAS Number:
Molecular Weight:
390.34
Products
Formula Weight
Manufacturer
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing