![Monoclonal Anti-Cytokeratin, pan−FITC antibody produced in mouse clone C-11, purified immunoglobulin, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/371/943/0ba7b1e3-8645-464a-990d-4fd08ae4ebba/640/0ba7b1e3-8645-464a-990d-4fd08ae4ebba.jpg)
Monoclonal Anti-Cytokeratin, pan−FITC antibody produced in mouse
![Monoclonal Anti-Cytokeratin, pan−FITC antibody produced in mouse clone C-11, purified immunoglobulin, buffered aqueous solution](/deepweb/assets/sigmaaldrich/product/images/371/943/0ba7b1e3-8645-464a-990d-4fd08ae4ebba/640/0ba7b1e3-8645-464a-990d-4fd08ae4ebba.jpg)
Synonym(s):
Monoclonal Anti-Pan Cytokeratin
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing