![Monoclonal Anti-CLIP1 antibody produced in mouse Prestige Antibodies® Powered by Atlas Antibodies, clone CL4851, purified immunoglobulin, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/888/364/120d4bc0-0341-4401-a9bd-6d4e67b82b8a/640/120d4bc0-0341-4401-a9bd-6d4e67b82b8a.jpg)
Monoclonal Anti-CLIP1 antibody produced in mouse
![Monoclonal Anti-CLIP1 antibody produced in mouse Prestige Antibodies® Powered by Atlas Antibodies, clone CL4851, purified immunoglobulin, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/888/364/120d4bc0-0341-4401-a9bd-6d4e67b82b8a/640/120d4bc0-0341-4401-a9bd-6d4e67b82b8a.jpg)
Synonym(s):
CLIP, CLIP-170, CLIP170, CYLN1, RSN
Products
Product Category
Brand
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing