![Monoclonal Anti-Alkaline Phosphatase, Human Placental antibody produced in mouse clone 8B6, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/153/053/60c0a4e2-ffe7-439d-ab4c-4f31dc1bcc78/640/60c0a4e2-ffe7-439d-ab4c-4f31dc1bcc78.jpg)
Monoclonal Anti-Alkaline Phosphatase, Human Placental antibody produced in mouse
![Monoclonal Anti-Alkaline Phosphatase, Human Placental antibody produced in mouse clone 8B6, ascites fluid](/deepweb/assets/sigmaaldrich/product/images/153/053/60c0a4e2-ffe7-439d-ab4c-4f31dc1bcc78/640/60c0a4e2-ffe7-439d-ab4c-4f31dc1bcc78.jpg)
Synonym(s):
Monoclonal Anti-Alkaline Phosphatase, Human Placental, Anti-Phosphatase, Alkaline, Human Placental, PLAP
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing