Skip to Content
Merck
Methyl valerate analytical standard

Methyl valerate

Methyl valerate analytical standard
Synonym(s):
Methyl pentanoate
Linear Formula:
CH3(CH2)3CO2CH3
CAS Number:
Molecular Weight:
116.16
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing