Methyl Purple
Synonym(s):
Methyl Purple indicator solution
Empirical Formula (Hill Notation):
C67H62N8Na4O14S3
CAS Number:
Molecular Weight:
1391.41
Products
Product Category
Brand
Formula Weight
pH Value
Physical Form
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing