![Methylphenobarbital British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/698/032/e88f968a-0116-49b1-81fc-bed3107c1bc6/640/e88f968a-0116-49b1-81fc-bed3107c1bc6.png)
Methylphenobarbital
![Methylphenobarbital British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/698/032/e88f968a-0116-49b1-81fc-bed3107c1bc6/640/e88f968a-0116-49b1-81fc-bed3107c1bc6.png)
Empirical Formula (Hill Notation):
C13H14N2O3
CAS Number:
Molecular Weight:
246.26
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2