![Methylhydroquinone analytical standard](/deepweb/assets/sigmaaldrich/product/structures/266/057/2c629f6c-78be-4f1b-a6e6-f76b450acde6/640/2c629f6c-78be-4f1b-a6e6-f76b450acde6.png)
Methylhydroquinone
![Methylhydroquinone analytical standard](/deepweb/assets/sigmaaldrich/product/structures/266/057/2c629f6c-78be-4f1b-a6e6-f76b450acde6/640/2c629f6c-78be-4f1b-a6e6-f76b450acde6.png)
Synonym(s):
Toluhydroquinone
Linear Formula:
CH3C6H3-1,4-(OH)2
CAS Number:
Molecular Weight:
124.14
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing