![Methyl 4-bromo-1-methyl-1H-pyrrole-2-carboxylate AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/349/662/83b409c1-7623-4041-9c32-151875bae9f9/640/83b409c1-7623-4041-9c32-151875bae9f9.png)
Methyl 4-bromo-1-methyl-1H-pyrrole-2-carboxylate
![Methyl 4-bromo-1-methyl-1H-pyrrole-2-carboxylate AldrichCPR](/deepweb/assets/sigmaaldrich/product/structures/349/662/83b409c1-7623-4041-9c32-151875bae9f9/640/83b409c1-7623-4041-9c32-151875bae9f9.png)
Empirical Formula (Hill Notation):
C7H8BrNO2
CAS Number:
Molecular Weight:
218.05
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing