Skip to Content
Merck
Methyl 2-furoate 98%

Methyl 2-furoate

Methyl 2-furoate 98%
Synonym(s):
Methyl furan-2-carboxylate, Methyl pyromucate
Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing