![Methiocarb certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/360/007/e6ff8175-dd88-49b4-87ab-ecfeef3fb2d8/640/e6ff8175-dd88-49b4-87ab-ecfeef3fb2d8.png)
Methiocarb
![Methiocarb certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/360/007/e6ff8175-dd88-49b4-87ab-ecfeef3fb2d8/640/e6ff8175-dd88-49b4-87ab-ecfeef3fb2d8.png)
Synonym(s):
Mercaptodimethur
Empirical Formula (Hill Notation):
C11H15NO2S
CAS Number:
Molecular Weight:
225.31
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2