![Meloxicam Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/824/931/0437cc72-29ba-4cda-8f7a-7497cfcb3db2/640/0437cc72-29ba-4cda-8f7a-7497cfcb3db2.jpg)
Meloxicam Related Compound C
![Meloxicam Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/824/931/0437cc72-29ba-4cda-8f7a-7497cfcb3db2/640/0437cc72-29ba-4cda-8f7a-7497cfcb3db2.jpg)
Synonym(s):
Isopropyl 4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxylate 1,1-dioxide
Empirical Formula (Hill Notation):
C13H15NO5S
CAS Number:
Molecular Weight:
297.33
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2