Skip to Content
Merck
Manganese(II) acetate tetrahydrate ≥99%

Manganese(II) acetate tetrahydrate

Manganese(II) acetate tetrahydrate ≥99%
Synonym(s):
Manganous acetate, Manganese(II)acetate tetrahydrate
Linear Formula:
(CH3COO)2Mn · 4H2O
CAS Number:
Molecular Weight:
245.09

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Core
Special Grade