![Lyticase from Arthrobacter Luteus Suitable for manufacturing of diagnostic kits and reagents, lyophilized powder, ≥2000 units/mg protein](/deepweb/assets/sigmaaldrich/product/images/317/547/8596e22b-3b57-4833-b5ba-f30fe0752dcf/640/8596e22b-3b57-4833-b5ba-f30fe0752dcf.jpg)
Lyticase from Arthrobacter luteus
![Lyticase from Arthrobacter Luteus Suitable for manufacturing of diagnostic kits and reagents, lyophilized powder, ≥2000 units/mg protein](/deepweb/assets/sigmaaldrich/product/images/317/547/8596e22b-3b57-4833-b5ba-f30fe0752dcf/640/8596e22b-3b57-4833-b5ba-f30fe0752dcf.jpg)
Synonym(s):
Lyticase from Arthrobacter luteus
CAS Number:
Products
Product Category
Brand
Biological Source
Physical Form
Chemical Composition
Quality Segment
Application
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing