Luzindole
Synonym(s):
N-Acetyl-2-benzyltryptamine, N-[2-[2-(Phenylmethyl)-1H-indol-3-yl]ethyl]acetamide
Empirical Formula (Hill Notation):
C19H20N2O
CAS Number:
Molecular Weight:
292.37
Products
Product Category
Brand
Formula Weight
Gene Alias
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing