Skip to Content
Merck
L-Rhamnose monohydrate analytical standard

L-Rhamnose monohydrate

L-Rhamnose monohydrate analytical standard
Synonym(s):
6-Deoxy-L-mannose, L(+)Rhamnopyranose
Empirical Formula (Hill Notation):
C6H12O5 · H2O
CAS Number:
Molecular Weight:
182.17

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Technique