Skip to Content
Merck
Losartan carboxylic acid ≥95% (HPLC)

Losartan carboxylic acid

Losartan carboxylic acid ≥95% (HPLC)
Synonym(s):
EXP3174, EXP-3174
Empirical Formula (Hill Notation):
C22H21ClN6O2
CAS Number:
Molecular Weight:
436.89

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing