Loratadine Impurity E
Synonym(s):
Ethyl 4-(13-chloro-4-azatricyclo[9.4.0.03,8]pentadeca-1(11),3(8),4,6,12,14-hexaen-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate
Empirical Formula (Hill Notation):
C22H23ClN2O2
CAS Number:
Molecular Weight:
382.88
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing