Lead(II) tetrafluoroborate solution
Synonym(s):
Lead boron fluoride, Lead(II) fluoroborate
Linear Formula:
Pb(BF4)2
CAS Number:
Molecular Weight:
380.81
Products
Brand
Formula Weight
Physical Form
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing