
Latanoprost Related Compound E

Synonym(s):
Latanoprost acid, (5Z,9α,11α,15R)-9,11,15-Trihydroxy-17-phenyl-18,19,20-trinor-prost-5-en-1-oic acid, (Z)-7-[(1R,2R,3R,5S)-3,5-Dihydroxy-2-[(3R)-3-hydroxy-5-phenylpentyl]cyclopentyl]hept-5-enoic acid, PhXA 85
Empirical Formula (Hill Notation):
C23H34O5
CAS Number:
Molecular Weight:
390.51
Products
Brand
Feature
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2