
Lamotrigine Related Compound C

Synonym(s):
3-Amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-5(2H)-one
Empirical Formula (Hill Notation):
C9H6Cl2N4O
CAS Number:
Molecular Weight:
257.08
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2