
Ir(MDQ)2(acac)

Synonym(s):
Bis(2-methyldibenzo[f,h]quinoxaline)(acetylacetonate)iridium(III)
Empirical Formula (Hill Notation):
C39H29N4O2Ir
CAS Number:
Molecular Weight:
777.89
Products
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing